EP0030127714
Eppendorf® Serological Pipets, Sterile Grade
blue, capacity 5 mL, pack of 400 ea (4 bags × 100 pcs.)
Scegli un formato
118,00 €
Scegli un formato
About This Item
118,00 €
Prodotti consigliati
Materiali
blue
Sterilità
sterile
Caratteristiche
DNA free
graduations
plugged
Confezionamento
pack of 400 ea (4 bags × 100 pcs.)
Produttore/marchio commerciale
Eppendorf® 0030127714 (Global)
Capacità
5 mL
Descrizione generale
- Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
- Easy-to-read graduations for accurate volume determination.
- Pipets are color-coded based on sizes for easy identification.
- Sterility assurance level of 10-6.
- Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
- Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.
Applicazioni
- Transfer of liquids
- Mixing suspensions
- Combining reagents and chemical solutions
- Transferring cells for empirical analysis or expansion
- Layering reagents for creating higher density gradients
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)