EP0030113535
Eppendorf® Deepwell 96 Well PCR Plate with SafeCode System (data matrix code, barcode and plain text labels)
1000 µL per well, clear white wells, PCR clean
About This Item
Materiali
clear white wells
polypropylene
Sterilità
non-sterile
Caratteristiche
PCR clean
lid: no
Confezionamento
pkg of 20 plates (5 bags × 4 plates)
Produttore/marchio commerciale
Eppendorf® 0030113535
Misura
96 well
Dimensione pozzetto
1000 μL , per well
Compatibilità
suitable for PCR
Categorie correlate
Descrizione generale
Caratteristiche e vantaggi
- 3-level coding for safe sample and vessel identification
- Codes on three sides of plates for flexible reading
- ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
- Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
- Direct import into eLabInventory and eLabJournal software for convenient handling
- Import to other database systems possible
- Manufactured from high-quality polypropylene for highest sample purity
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)