EP0030113527
Eppendorf® Deepwell 96 Well PCR Plate with SafeCode System (data matrix code, barcode and plain text labels)
2000 µL per well, clear white wells, PCR clean
About This Item
Codice UNSPSC:
41106300
NACRES:
NB.15
Prodotti consigliati
Materiali
clear white wells
polypropylene
Sterilità
non-sterile
Caratteristiche
PCR clean
lid: no
Confezionamento
pkg of 20 plates (5 bags × 4 plates)
Produttore/marchio commerciale
Eppendorf® 0030113527
Misura
96 well
Dimensione pozzetto
2000 μL , per well
Compatibilità
suitable for PCR
Descrizione generale
Deepwell Plate 96/2000 µL, wells clear, PCR clean, white, with 2D SafeCode (DataMatrix/ECC200) and human readable code, 20 plates (5 bags x 4 plates)
Caratteristiche e vantaggi
- 3-level coding for safe sample and vessel identification
- Codes on three sides of plates for flexible reading
- ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
- Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
- Direct import into eLabInventory and eLabJournal software for convenient handling
- Import to other database systems possible
- Manufactured from high-quality polypropylene for highest sample purity
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)