Documenti
CLS7095D9
Corning® disposable Pasteur pipettes, bulk pack, non-sterile, unplugged
size 9 in., borosilicate
Sinonimo/i:
Pipettes, pipettes, serological pipets, serological pipettes
About This Item
Codice UNSPSC:
41121500
eCl@ss:
32079210
NACRES:
NB.04
Materiali
borosilicate glass
Sterilità
non-sterile
Confezionamento
pack of 200 ea
Produttore/marchio commerciale
Corning 7095D-9
Misura
9 in.
Descrizione generale
  • Corning® disposable Pasteur pipettes are made of sturdy and optically clear borosilicate glass.
  • Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
  • Feature a constriction one inch below the top to facilitate plugging.
  • Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
Note legali
Corning is a registered trademark of Corning, Inc.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 3
1 of 3
Pipette Pasteur Sigma cotton plugged, L 5 3/4 in.
S5893
Pipette Pasteur Sigma
BRAND® dropping pipettes capacity 5 mL, with integrated bellows
Z331805
BRAND® dropping pipettes