Documenti fondamentali
CLS431220
Corning® syringe filters
cellulose acetate membrane (surfactant-free), diam. 28 mm, pore size 0.45 μm
Sinonimo/i:
disposable syringe filters, sterile syringe filters, syringe filters
Scegli un formato
50 EA
180,00 €
180,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
50 EA
180,00 €
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
180,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
acrylic copolymer housing
cellulose acetate membrane (surfactant-free)
Sterilità
sterile
Confezionamento
case of 50
Produttore/marchio commerciale
Corning 431220
Diametro
28 mm
Dimensione pori
0.45 μm
Raccordi
female Luer inlet (lock)
male Luer outlet (slip)
Descrizione generale
Corning syringe filters are 100% integrity tested and are certified nonpyrogenic and noncytotoxic and are manufacturered in accordance with ISO 9002 standards. A variety of membranes are available to meet your needs.
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
These Corning syringe filters have acrylic copolymer housings, female luer lock inlets and male luer slip outlets.
  • SFCA-PF (surfactant-free cellulose acetate with prefilter) membranes provide the lowest protein binding and have an integral glass fiber prefilter for extra filtering capacity
  • 100% integrity tested, sterile and certified nonpyrogenic and noncytotoxic
  • Manufactured in accordance with ISO 9002 standards
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Qiushi Wei et al.
International journal of molecular medicine, 45(3), 816-824 (2020-01-28)
Icaritin, a metabolite of icariin, is a potent promoter of bone marrow‑derived mesenchymal stem cells (BMSCs) osteogenesis, but the underlying mechanisms remain unclear. To examine the effects of icaritin on osteogenic differentiation, BMSCs were exposed to osteogenic induction medium with