Documenti fondamentali
CLS3830
Corning® spheroid microplates
384 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile
Sinonimo/i:
3D cell culture plates, Spheroid culture plates
Scegli un formato
50 EA
2.660,00 €
2.660,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
50 EA
2.660,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.24
2.660,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
clear bottom
polystyrene
round black/clear bottom
Sterilità
sterile
Caratteristiche
lid
skirt
Specific plates for 3D cell culture
ULA (Ultra-Low Attachment surface)
Confezionamento
pkg of 10 ea
pkg of 50 ea
Produttore/marchio commerciale
Corning 3830
Misura
384 wells
Volume pozzetto
90 μL
Pozzetti
384 wells
Volume d’esercizio
20-80 μL
Compatibilità
suitable for (cell cuture)
Tipo di binding
(ultra-low attachment)
Descrizione generale
Key Features:
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel proprietary well geometry aids formation of spheroids in center of well
  • Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
  • Uniform, single spheroid formation across all wells
  • Centering of the spheroid in each well enables automated visualization
  • Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
  • Ease of media exchange or treatment with test compounds/drugs
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
Discover how Corning® Spheroid Microplates played a key role in the use of use of 3D cell cultures for in vitro drug discovery assay.
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models