Vai a
B9907
Nalgene® wide mouth Lab Quality PPCO bottle
capacity 250 mL, translucent, with cap, pack of 12 ea
Sinonimo/i:
nalgene bottle, nalgene bottles, Nalgene 2105-0008, laboratory container, receptical, vessel
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
A voi/SKUDisponibilitàPrezzo
12 ea
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
42,30 €
Informazioni su questo articolo
NACRES:
NB.17
UNSPSC Code:
41121800
42,30 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
polypropylene cap, polypropylene copolymer bottle, polypropylene copolymer bottles (PPCO), round bottle
joint
Joints threaded neck (43)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2105-0008
bottle diam. × H
61 mm × 131 mm
capacity
250 mL
color
translucent
General description
Store specimens where autoclaving is required is these Nalgene® Wide-Mouth Lab Quality PPCO Bottles. Made from high-quality, laboratory-grade plastic materials for reliably low leachables and extractables.
Features and Benefits
  • Excellent durability and are chemically resistant to most acids, bases, and alcohols.
  • Laboratory-grade PPCO material for reliably low leachables and extractables.
  • Translucent - for visualization of samples.
  • A wide neck enables easy transferring of contents.
  • Linerless caps ensure a leak-proof seal for sample protection.
  • Autoclavable - The bottles can withstand temperatures up to +121°C.
  • Manufactured from non-toxic materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
B9782B9657B9532
description
capacity 250 mL, translucent, with cap, pack of 12 ea
description
capacity 125 mL, translucent, with cap, pack of 12 ea
description
capacity 60 mL, translucent, with cap, pack of 12 ea
description
capacity 30 mL, translucent, with cap, pack of 12 ea
manufacturer/tradename
Nalgene® 2105-0008
manufacturer/tradename
Nalgene® 2105-0004
manufacturer/tradename
Nalgene® 2105-0002
manufacturer/tradename
Nalgene® 2105-0001
feature
with cap
feature
with cap
feature
with cap
feature
with cap
capacity
250 mL
capacity
125 mL
capacity
60 mL
capacity
30 mL
material
round bottle, polypropylene copolymer bottles (PPCO), polypropylene cap
material
round bottle, polypropylene cap, polypropylene copolymer bottles (PPCO)
material
polypropylene copolymer bottle, polypropylene copolymer bottles (PPCO), round bottle, polypropylene cap
material
polypropylene cap, polypropylene copolymer bottle, polypropylene copolymer bottles (PPCO), round bottle
joint
Joints threaded neck (43)
joint
Joints threaded neck (28)
joint
Joints threaded neck (28)
joint
Joints threaded neck (28)
bottle diam. × H
61 mm × 131 mm
bottle diam. × H
50 mm × 90 mm
bottle diam. × H
39 mm × 86 mm
bottle diam. × H
34 mm × 63 mm
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)