Documenti fondamentali
AXYP5ML48C
Deep well plate, with pyramid bottom
48 rectangular wells, pyramid bottom clear polypropylene wells, maximum volume 5 mL, 25 ea of
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
pyramid bottom clear polypropylene wells
Sterilità
non-sterile
Caratteristiche
lid: no
skirt
Confezionamento
case of 25 ea
pack of 5 ea
25 ea of
Produttore/marchio commerciale
Axygen Inc P-5ML-48-C
Corning
Volume massimo
5 mL
5.0 mL
Misura
48 wells
Volume d’esercizio
4.6 mL
Tipo di binding
non-treated surface