Documenti fondamentali
Z758027
BRAND® PLT unit (pipette leak testing unit)
AC/DC input 100 - 240 V AC
Sinonimo/i:
accuracy of a pipette, calibrating pipettes, pipette accuracy
Scegli un formato
1 EA
1.410,00 €
1.410,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
1.410,00 €
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
1.410,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
BRAND 703970
Categorie correlate
Descrizione generale
Improve process reliability with functional validation of air-displacement pipettes!
The most frequent cause of inaccuracy in piston-operated pipettes is leakage. This arises from damage either to the seals, pistons, or tip cones. Often not detectable by the naked eye, leaks lead to significant volume errors. The BRAND pipette leak tester (PLT unit) for air displacement pipettes finds even the smallest leaks within seconds.
  • Limit values for the commercially available single- and multichannel pipettes in the volume range 1 μL to 10 mL are pre-programmed
  • Testing with and without tips
  • Test results in seconds
  • Patent pending
While the PLT unit cannot replace regular gravimetric testing, daily pipette checks can provide a safeguard during the periods between calibrations. Even the smallest leaks are detected! Process reliability for the pipettes is thus significantly improved.
Unit includes: one adapter each for testing single-channel air-displacement pipettes with tip (mounted) and without tip, 2 plugs (for instrument validation), 3 replacement PE filters for the pipette adapters, universal AC adapter, quality certificate and operating manual.
Caratteristiche e vantaggi
  • Functional testing of your air-displacement pipettes in seconds.
  • Automatic documentation of test results with the PLTconnect software.
  • More security while pipetting.
Altre note
BR703970 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)