Documenti fondamentali
Z742348
2MAG Asteroid extra strength stirring bar
diam. × L 28 mm × 40 mm
Sinonimo/i:
magentic flea, stirbar
Scegli un formato
1 EA
58,00 €
58,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
58,00 €
About This Item
Codice UNSPSC:
41103800
NACRES:
NB.72
58,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
PTFE cover
interior (SmCo)
triangular
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
2mag 44040
diam. × lung.
28 mm × 40 mm
Peso
0.06 kg
Descrizione generale
New developed, high-effective stirring bar, ideal for large mixing volumes, viscous media, stirring over wide distances or for disadvantageous vessel bottoms.
More than 4x higher torque transmissible, unbeatable increased mixing effect in comparison to conventional stirring bars, high energetic and long-term stable magnetic field, no demagnetization effect, concave design for a minimum of bearing surface and reliable rotation, triangular shape for powerful shear force, better mixing effect at lower rotation speed.
300 percent less PTFE abrasion than comparable round shape stirring bars, significantly higher rotation speed reachable, high quality, even and sealed PTFE cover, FDA-conform, steam sterilizable at 121 degrees C.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)