Documenti fondamentali
Z742327
2MAG maxMIX single point stirrers up to 150 L,
AC/DC input 240 V AC, UK plug
Sinonimo/i:
high volume stirrer, induction plate, induction stirrer, laboratory stirrer, magnetic stir plate, magnetic stirrer
Scegli un formato
1 EA
2.910,00 €
2.910,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
2.910,00 €
About This Item
Codice UNSPSC:
41103800
NACRES:
NB.72
2.910,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
stainless steel housing
stainless steel plate
Caratteristiche
positions 1
40 W Stirring Power (
4 step)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
2mag 30501
Parametri
-10-50 °C temp. limit (at 80 percent humidity)
100-1200 rpm speed
Largh. × prof. × alt.
265 mm × 320 mm × 68 mm
Dimensioni della piastra × Largh. × Lungh.
× 260 mm × 290 mm
Volume max.
1-150 L , per stirring position
Peso
~9.3 kg
Descrizione generale
Unbeaten strong magnetic stirrer with large set-up area, up to 150 litres stirring volume, also suitable for highly viscous media.
Very strong magnetic coupling, therefore predestined for magnetic power transmission over wide distances (up to 60 mm), for thick vessel bottoms or for double-coat vessels. Maintenance-free 2mag-motor-drive with large speed range from 100 up to 1,200 rpm, fully encapsulated and tight stainless steel housing, IP64, extremely robust construction and performance, easy to clean under running water and firm, individual measurements on request. Integrated control unit with strong 40 watts stirring power,large speed range from 100 up to 1,200 rpm, SoftStart-function (variable 20-99 seconds) for reliable catching/centring and safe accelerating of the stirring bar at different volumes and viscosities.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)