Documenti fondamentali
Z740177
Millipore
Amicon® Ultra-0.5
Ultracel-50 membrane, 50 kDa
About This Item
Codice UNSPSC:
41104816
Materiali
regenerated cellulose membrane
styrene-butadiene housing
Sterilità
non-sterile
Caratteristiche
50 kDa
Confezionamento
pack of 24 ea
Produttore/marchio commerciale
Millipore UFC505024
Dimens. tubo
~1.5 mL
Volume d’esercizio
0.5 mL
Dimensione pori
50 kDa MWCO
Note legali
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Esteban Cruz et al.
Cancers, 11(6) (2019-06-27)
Nanoparticle carriers offer the possibility of enhanced delivery of therapeutic payloads in tumor tissues due to tumor-selective accumulation through the enhanced permeability and retention effect (EPR). Gold nanoparticles (AuNP), in particular, possess highly appealing features for development as nanomedicines, such