Documenti fondamentali
Z693537
2mag bioMIXcontrol MS4 control unit
AC/DC input 230 V AC, Schuko plug
Sinonimo/i:
laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
Scegli un formato
1 EA
2.730,00 €
2.730,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
2.730,00 €
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
2.730,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
stainless steel plate
Caratteristiche
CE compliant
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
2mag 98604
Parametri
5 to 250 rpm speed (stirring)
5-250 rpm speed
Dimensioni della piastra × Largh. × Lungh.
× 200 mm × 110 mm
Capacità
capacity (4 x bioMIXdrive 1)
Descrizione generale
Efficient control unit, for 4 drives 2mag bioMIXdrive 1, developed for the hard daily lab use and warming-free continuous operation.
Control unit for the connection of 4 stirring drive bioMIXdrive 1, individual and independent speed setting for each stirring drive (Master-Slave-Technology), also ON/OFF function, synchronous speed mode for all drives, extended speed range from 5 to 250 rpm, 6 watts/stirring point without any own-generated heating effect, automatic power setting for the warming-free continuous operation e.g. in CO2-incubators, jerk-free stirring, even and smooth stirring also at slow speeds.
Digital display for speed setting, SoftStart for reliable and safe accelerating of stirring bar, QuickSet for quick setting of start- and maximum speed, storage of last working status when turning off, firm stainless steel housing, easy to clean.
Interfaces: 0-10 volts, 4-20 mA, RS 232, USB, Ethernet.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)