Documenti fondamentali
Z685232
Plas-Labs automatic anaerobic chamber AC input 230 V
AC/DC input 230 V AC, European 2-pin plug
Scegli un formato
1 EA
24.750,00 €
24.750,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
24.750,00 €
About This Item
Codice UNSPSC:
41103406
NACRES:
NB.83
24.750,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Descrizione
includes glove box, shelving unit and support cart
Produttore/marchio commerciale
Plas-Labs Simplicity-888
Descrizione generale
The Plas-Labs automatic anaerobic chamber has a single station which is designed for automatic atmosphere control when working with oxygen sensitive materials, product containment, and general isolation control. This chamber features an internal PC with multifunctional operator interface.
  • Three drying train canisters with quick disconnects
  • Programmable catalyst heater unit
  • Bright fluorescent light system
  • White ambidextrous Hypalon® gloves
  • Two 1/4 in. NPT gas connections
  • Two ground key cock valves on main chamber
  • Electrical outlet strip; UL & CSA approved, hospital grade with four outlets
  • CE compliant
Altre note
Price includes shipping
Note legali
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)