Documenti fondamentali
Z677655
Butoject butyl gloves
size L
Sinonimo/i:
chemically resistant glove
Scegli un formato
1 PAIR
50,80 €
50,80 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 PAIR
50,80 €
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
50,80 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
black butyl rubber
black gloves
Misura
L
Sterilità
non-sterile
Caratteristiche
CE compliant
latex free
powder-free: no
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Confezionamento
pack of 1 pair
Produttore/marchio commerciale
KCL 897
Lungh. × spessore
360 mm × 0.3 mm
Descrizione generale
Butoject is a highly flexible and effective chemical protective glove made of Butyl rubber, suitable for extreme demands for work with ester′s and ketone′s. With a thickness of merely 0.30 mm and very good cold flexibility
Butoject is very comfortable to wear and allows good dexterity. To improve the grip on smooth and wet parts, this glove has an additionally roughened surface.
Caratteristiche e vantaggi
  • Provides excellentprotection against aggressive chemicals, helping lab technicians work safely with harsh substances.
  • Ensures wearing comfort and dexterity due to its high elasticity and ergonomic design, reducing fatigue during long tasks.
  • Improved grip on wetsurfaces with a roughened surface, enhancing safety and handling of equipment.
  • Made of durable butyl rubber, offering good mechanical resistance and extended usability, reducing the need for frequent replacement.
  • Standard type EN 374 (cat.III) and EN 388 (cat. III) compliant, ensuring a high level of chemical and mechanical protection.
  • Powder-free design minimizes contamination and is CE compliant for safe use in laboratory environments.
  • Reusable, making it cost-effective for labs with frequent glove usage.
  • 360 mm length and 0.3 mmthickness provide extensive coverage while maintaining flexibility and coldresistance.
Note legali
Butoject is a trademark of KCL GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)