Documenti fondamentali
Z630020
IKA® HS 501 Horizontal shaker
AC/DC input 230 V AC
Sinonimo/i:
lab shaker, laboratory shaker, orbital shaker, reciprocating shaker
Scegli un formato
1 EA
5.660,00 €
1 EA
5.660,00 €
1 EA
5.660,00 €
5.660,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
5.660,00 €
1 EA
5.660,00 €
1 EA
5.660,00 €
About This Item
Codice UNSPSC:
41103811
NACRES:
NB.73
5.660,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Caratteristiche
CE compliant
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
IKA 0025002982 (Europe) and 0025004443 (GB)
Descrizione generale
comes with integrated USD and RS 232 interface
Low profile laboratory shaker with a pleasant design, large mounting surface and a loading capacity of 33 lbs., depending on the attachment. The horizontal shaker features a reciprocating motion. Infinitely variable speed control from 0-300 rpm, digitally displayed. Timer 0-56 min and setting for continuous operating. Ideally suitable for vessels with a volume of more than 250 mL, e.g. round flasks, Erlenmeyer flasks, culture flasks, culture bottles.
For biological and microbiological growth tests, for the production of stock solution, etc.
Guaranteed continuous operation even under extreme loads.
Note legali
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)