Z531626
Tubing Adapters with Torion™ Connection
O.D. 3/16 in., Straight, male joint: ST/NS 14/20
Scegli un formato
93,00 €
Scegli un formato
About This Item
93,00 €
Prodotti consigliati
Giunto
male joint: ST/NS14/20
D.E.
3/16 in.
4.8 mm
Categorie correlate
Descrizione generale
5/16 in. O.D fits a #8 Torion, 1/2 in. O.D. fits a #14 Torion, and 3/16 in. O.D fits a #5 Torion.
- Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE-coated sealing ring of the adapter
- Turn the bushing to make an airtight compression seal with the tubing
- Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use
- O.D. refers to the tubing outer diameter
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)