Documenti
Z303348
Quickfit® double surface (Davies) condenser
L 150 mm, joint: ST/NS 14/23
Sinonimo/i:
C5/11/SC
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Giunto
joint: ST/NS14/23
Produttore/marchio commerciale
Quickfit C 5/11/SC
Disponibilità
not available in USA and Canada
not available in USA and Canada
Lunghezza
150 mm
Area superficiale
170 cm2
Descrizione generale
Increased surface area and effectiveness when compared with the corresponding Liebig condenser. Inner cooling surface gives a baffling effect, while outer cooling surface prevents creep and loss of vapour when used with low boiling liquids.
Note legali
Quickfit is a registered trademark of SciLabware Limited