Documenti
Z303070
Quickfit® multi-limb vacuum receiver
Quickfit RA 12/2, sloping delivery adapter
Sinonimo/i:
RA12/2
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Giunto
female joint 19/26
male joint 19/26
Produttore/marchio commerciale
Quickfit RA 12/2
Disponibilità
not available in ROW (Available only in EU countries)
not available in USA and Canada
Descrizione generale
When suitably lubricated the receiver adapter may be rotated at the conical joint to divert liquid, in turn, to four attached flasks without disturbing the vacuum in the system. Requires four 100 mL flasks (Z302619, not included) with screw-thread adapter for flexible tubing.
Note legali
Quickfit is a registered trademark of SciLabware Limited