Documenti fondamentali
Z270334
Sigma-Aldrich
Bunsen burner
Natural gas: Meker
Sinonimo/i:
bunsen burner gas, bunsen burner lab, bunsen burners
About This Item
Codice UNSPSC:
41100000
Descrizione generale
For Natural Gas. For high temperature work. Nickel plated brass tube with rotatable non-removable air regulator. Perforated metal burner head 25 mm diameter produces an evenly aerated flame of uniform intensity. On heavy stable cast iron base, baked hammertone finish, with serrated gas inlet tube 9 mm diameter.
Laboratory gas burners for use with natural (methane) or cylinder (propane) gases.
Available in 3 styles:
  • General-purpose - for general laboratory applications (solid base)
  • Fast-flame - with flame stabilizer to control flame temperature and length (three spoke base)
  • Meker - for applications where an extremely hot flame is required (only for bottled gas, perforated metal mesh burner)
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)