Documenti
Z175552
Sigma-Aldrich
Lecture-bottle valve
CGA Inlet for 180M/110F, brass
Sinonimo/i:
Lecture Bottle Control Valve
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Materiali
brass
Livello qualitativo
Raccordi
CGA Inlet for 180M/110F
Descrizione generale
Diffusion resistant, diaphragm seal, packless on/off valve. 3/8 in. NPTM valve thread. CGA 180M inlet/110F outlet connection.