Documenti
Z146951
Sigma-Aldrich
Lecture-bottle control valve
stainless steel, hose outlet I.D. 1/4 in.
Sinonimo/i:
Stainless steel lecture-bottle control valve
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Materiali
stainless steel
Giunto
joint CGA Inlet
joint hose outlet
Parametri
-65-250 °F temperature (-54-120 °C)
3,000 psi pressure range (@ 70°F)
Diam. int. uscita tubo
1/4 in.
Raccordi
CGA 110 male
Descrizione generale
CGA110, straight-pattern valve for precise flow control of lecture-bottle gas. Requires CGA110 PTFE sealing washer.
Brass control valve shown.
Attenzione
Gas-control valves do not control pressure in a closed system. Regulator or relief valves should be used to prevent the buildup of unsafe pressures that could lead to explosion.