Z168572
Thermolyne® Cimarec®-top stirring hotplate
AC/DC input 120 V AC, plate size 7 in. × 7 in.
About This Item
Codice UNSPSC:
41102406
Prodotti consigliati
Sterilità
non-sterile
Caratteristiche
CSA approved
Produttore/marchio commerciale
Barnstead International SP46925
Disponibilità
not available in EU
Dimens. piastra
7 in. × 7 in.
Uscita
118 watts
Descrizione generale
Has all features of the Cimarec-top hot plate plus has excellent low-speed stirring control and speed range of 60 to 1,000 rpm. All units are CSA listed.
Note legali
Cimarec is a registered trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)