Documenti fondamentali
WHAWB100024
Qiagen® Multi Barrier Pouch
large, size , L x W 7" x 7.37" (17.8 cm x 18.7 cm), resealable, pkg of 50 ea
Sinonimo/i:
Multi Barrier Pouch Reseal 7 x 7.37 (50), Whatman FTA, Z747351
Scegli un formato
1 PKG
205,00 €
205,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 PKG
205,00 €
About This Item
Codice UNSPSC:
41105500
NACRES:
NB.22
205,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Confezionamento
pkg of 50 ea
Produttore/marchio commerciale
Qiagen WB100024
Misura
, L x W 7" x 7.37" (17.8 cm x 18.7 cm)
Descrizione generale
Large size 7 x 7.37 inch (17.8 x 18.7 cm) resealable Multi-Barrier Pouches for transport or storage of QIAcard FTA CloneSaver.
Caratteristiche e vantaggi
  • Laminated layers maintain sample integrity and security by protecting the card from exposure to gas or liquid contamination.
  • The outer paper surface is suitable for labeling or writing. The Multi-Barrier Pouch has a resealable closure. To optimize sample integrity FTA Cards are stored in a Multi-Barrier Pouch with a desiccant packet.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Articoli
Description of common problems that can occur during genomic DNA preparation
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
Protocolli
A number of options are available for sample collection, transport, archiving, and DNA purification.
Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.