Documenti fondamentali
WHA69712504
Whatman® GD/XP syringe filters
Nylon membrane
Sinonimo/i:
Z754684
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Materiali
Nylon membrane
polypropylene housing
Sterilità
non-sterile
Confezionamento
pk of 1500
Produttore/marchio commerciale
Whatman 6971-2504
Whatman Article No. 28420562 (US reference)
Diametro
25 mm
Misura
Volume
≤25 mL
Dimensione pori
0.45 μm
Raccordi
female Luer inlet (lock)
male Luer outlet
Caratteristiche e vantaggi
  • Incorporates a two-layer polypropylene prefilter stack rather than a multi-layer glass fiber stack to minimize levels of ion extractables
  • Extremely low protein binding syringe filter
  • High loading volume for filtration of challenging samples
Compatibilità
Suitable for filtering trace metal analysis samples and for particulate laden sample preparation.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)