Documenti fondamentali
TEM-FCF400NIUA
Sigma-Aldrich
Ultrathin Formvar/Carbon Supported Nickel Square Mesh
size 400 mesh, box of 50
Sinonimo/i:
FOMVAR/CARBON MESH - NI 400 MESH UA 50/BX
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 50
Misura
400 mesh
Descrizione generale
Ultrathin 5-6nm Formvar and 1nm Carbon Supported Square Mesh Grids: Nickel Grid, 400 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)