Documenti fondamentali
TEM-7435702
Sigma-Aldrich
Lacey Carbon Supported Copper Tomography Grid R14C2
pack of 50 ×
Sinonimo/i:
Lacey Carbon on Tomography Grid, 50pk
About This Item
Codice UNSPSC:
41111761
Confezionamento
pack of 50 ×
Descrizione generale
This 1.5mm square, 300 mesh grid is made from copper, supported by lacey carbon, and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)