The majority of Synthware glass products, including Product SYNS322044, are made of Class A Borosilicate glass with 33 expansion. Unfortunately, they do not offer anything in Pyrex.
SYNS322044
Synthware™ straight bore glass stopcock
bore size 6 mm, O.D. 12 mm stem, plug size 20/44
Sinonimo/i:
Synthware S322044, two way straigh bore stopcock
Scegli un formato
68,80 €
Scegli un formato
About This Item
68,80 €
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S322044
Dimensioni foro
6 mm
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
-
What is the nature of the glass used for Product SYNS322044?
1 risposta-