Documenti
SYNF773030C
Synthware Gooch low form crucible
30 mL, frit O.D. 30 mm, porosity: coarse
Sinonimo/i:
Synthware F773030C, Gooch filter
About This Item
Codice UNSPSC:
41122200
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F773030C
Diam. est. frit
30 mm
Capacità
30 mL
Note legali
Synthware is a trademark of Kemtech America Inc.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 1
1 of 1
BRAND® filter crucible borosilicate glass, capacity 30 mL, 1 D 1
BR458021
BRAND® filter crucible