Documenti
SYNF621125G
Synthware pressure equalizing addition funnel
125 mL, joint: ST/NS 14/20, stopcock: glass
Sinonimo/i:
Synthware F621125G, dropping funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F621125G
Capacità
125 mL
125 mL
Descrizione generale
With glass stopcock, graduated.
Note legali
Synthware is a trademark of Kemtech America Inc.