SYNC244500
Synthware™ Allihn reflux condenser
joint: ST/NS 45/50, jacket L 260 mm
Sinonimo/i:
Synthware C244500, Soxhlet extraction condenser
About This Item
Prodotti consigliati
Giunto
joint: ST/NS45/50
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C244500
Lunghezza camicia
260 mm
Descrizione generale
Allihn reflux condenser with a water cooled inner drip joint at bottom.
Note legali
Synthware is a trademark of Kemtech America Inc.