SLW1132/15
Pyrex® Erlenmeyer flask, graduated, ground socket
joint: 29/32, capacity 250 mL
Sinonimo/i:
Pyrex, SciLabware 1132/15
Scegli un formato
290,00 €
Scegli un formato
About This Item
290,00 €
Prodotti consigliati
Materiali
borosilicate glass
flat bottom
Giunto
joint neck29/32
joint: 29/32
Descrizione
ground joint
Sterilità
non-sterile
Caratteristiche
graduated
without cap
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
SciLabware 1132/15
Capacità
250 mL
Diametro
85 mm
Descrizione generale
- Erlenmeyer flasks with high quality Quickfit® ground glass socket
- Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
Caratteristiche e vantaggi
- Complies with ISO 1773 and 4797
- Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
- Easy-to-read white enamel marking spot and graduated scale allows the approximate measurement of the flask contents
- N.B. Conical shape flasks are not suitable for use under vacuum, unless specifically engineered to do so.
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)