Documenti fondamentali
DWKW224585
WHEATON® clear sample vial with PTFE faced rubber lined cap packed in partitioned tray
glass, tube capacity (12 mL), screw cap
Sinonimo/i:
Clear Sample Vial, PTFE-Lined Vial, Sample Vial, W224585
Scegli un formato
144 EA
242,00 €
242,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
144 EA
242,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
242,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
flat bottom
glass
solid black phenolic cap
transparent borosilicate glass tube
Sterilità
non-sterile
Caratteristiche
with cap
Confezionamento
case of 144 ea partitioned tray
Produttore/marchio commerciale
WHEATON® W224585
Capacità
(12 mL)
Diam. tubo
(19 mm)
Raccordi
neck (External Thread)
Descrizione generale
Sample Vials in Lab File (With Caps Attached). Attached caps help maintain cleanliness. Supplied in partitioned trays to provide an easy way to store empty vials or to inventory samples.
  • Screw caps are attached to vials.
  • It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples, and metal foil liner for hydrocarbons.
  • Vials and caps are autoclavable.
  • 2 mL vials fit the M-T Vial File, 228778.
  • 4 mL vials fit the M-T Vial File, 228780.
  • Vials with caps attached packaged in partitioned trays
Caratteristiche e vantaggi
  • Clear vials are converted from Type I borosilicate glass tubing
  • Amber vials are converted from amber borosilicate glass tubing that conforms to USP Type I requirements for light transmission to protect light sensitive products
  • Sample vials with caps attached to vial help maintain cleanliness
  • Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • Shorty Vial® feature a low profile for greater stability and less dead volume
  • 14B rubber lined black phenolic screw cap for aqueous samples
  • PTFE / 14B rubber lined black phenolic screw cap for organic samples and improved chemical compatibility
  • Replacement caps can be purchased separately
Note legali
Lab File is a registered trademark of Wheaton Industries, Inc.
Shorty Vial is a registered trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)