This is a product of DWK Life Sciences under supplier product number 92812G-100. The manufacture does not provide the full product dimensions or product drawings.
Please see the manufacture website for more details on this product : https://www.dwk.com/na/kimble-kimax-volumetric-flask-class-a-heavy-duty-wide-mouth-glass-stopper-100-ml-92812g-100
DWK92812G-100
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask, glass
glass stopper, glass flask, flask capacity (100 mL), class A
Scegli un formato
392,00 €
Scegli un formato
About This Item
392,00 €
Prodotti consigliati
Materiali
glass flask
glass stopper
Caratteristiche
class A
Confezionamento
case of 6 ea
Produttore/marchio commerciale
Kimble® 92812G-100
Capacità pallone
(100 mL)
Descrizione generale
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Caratteristiche e vantaggi
- Manufactured from borosilicate glass 3.3
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Heavy duty walls provide superior strength, durability and safety
- Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Printed large volume numbers allow quick identification when selecting glassware
- Flat interior bottoms for stirring with standard stir bars
- Calibrate to contain
- Graduated to Class A volumetric tolerances
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
-
About this 100ml flask, could you tell us what is the width of the stem?
1 risposta-