Documenti fondamentali
CLS999075
Pyrex® ribbed watch glasses
O.D. ~75 mm
Sinonimo/i:
Corning® 9990
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.43
Produttore/marchio commerciale
Corning 9990-75
D.E.
~75 mm
Categorie correlate
Descrizione generale
Molded-in ribs allow vapors to escape during boiling to eliminate pressure buildup. Meets EPA requirements for methods 3005, 3010, 3020.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)