BR81167
BRAND® burette lateral stopcock
for burette capacity 25+50 mL, with spare key, PTFE key
Sinonimo/i:
Accessories, Burette stopcock, Burettes, Stopcocks
Scegli un formato
33,70 €
Scegli un formato
About This Item
33,70 €
Prodotti consigliati
Materiali
PTFE key
amber
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 81167
Categorie correlate
Descrizione generale
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)