Documenti fondamentali
BAF424002021
Scienceware® Lab Companion round vacuum desiccator
volume 6 L, clear polycarbonate, pack of 1 ea
Sinonimo/i:
Desiccator, Desiccator cabinet, Vacuum Bell Jar, Vacuum Desiccator
Scegli un formato
1 EA
393,00 €
393,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
393,00 €
About This Item
Codice UNSPSC:
41104510
NACRES:
NB.43
393,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
clear polycarbonate
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F42400-2021
Volume
6 L
6000 mL
Descrizione generale
These high-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind. Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods. A Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange.
Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
Available in both cabinet and round styles, all models include a desiccant tray (desiccants sold separately).
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • Locking ring secures top and bottom halves when vacuum not in use; provides additional convenience and safety when moving the unit
  • Six models to choose from; 6, 10, and 20 L internal volume in clear or UV blocking amber
Note legali
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Domande
  1. ¿Me podrían indicar las dimensiones de este desecador? Concretamente, el diámetro de la bandeja (y si viene o no incluida con el mismo) y la altura de las 2 partes útiles, es decir, la base desde la bandeja hasta la zona superior y la tapa (sin manómetro)
    1 risposta
    1. The supplier gives the Exterior Dimensions (W X H) as 28.6 x 35.4cm (11¼ x 13⅞”). The supplier also states that the item comes with 1 tray that can hold up to 15kg (33lb). They do not provide the dimensions of the tray. Please refer to the supplier data sheet:
      https://www.belart.com/media/catalogstudio/FAQS/Lab%20Companion%20Vacuum%20Desiccators.pdf