1681845
USP
Triclocarban Related Compound C
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1,3-Bis(3,4-dichlorophenyl)urea, 3,3′,4,4′-Tetrachlorocarbanilide
Select a Size
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
triclocarban
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Clc1c(ccc(c1)NC(=O)Nc2cc(c(cc2)Cl)Cl)Cl
InChI
1S/C13H8Cl4N2O/c14-9-3-1-7(5-11(9)16)18-13(20)19-8-2-4-10(15)12(17)6-8/h1-6H,(H2,18,19,20)
InChI key
ZDPIZPXVHVYTOK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service