Skip to Content
Merck
All Photos(1)

Key Documents

G005680

HPLC Analysis of Biogenic Amines on Ascentis® RP-Amide

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 5 μm particles (565305-U)
mobile phase: [A] 0.1% ammonium acetate in water (pH unadjusted); [B] acetonitrile
gradient: 25 to 75% B in 10 min; held at 75% B for 5 min
flow rate: 0.5 mL/min
column temp.: 35 °C
detector: UV, 340 nm
injection: 10 μL

suitability

application for HPLC

application(s)

clinical

Analysis Note

Following pre-column derivatization of primary amino groups using o-phthaldialdehyde/thiol reagent, the Ascentis RP-Amide was used for the analysis of biogenic amines commonly found in wines. These include histamine, spermine, spermidine, tyramine, putrescine and cadaverine. Structures along with an optimized chromatogram are presented.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Histamine
    Spermine
    Spermidine
    Tyramine
    1,4-Diaminobutane
    Cadaverine

analytical column

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service