Skip to Content
Merck
All Photos(1)

Key Documents

95065

Supelco

Terbuthylazine Metabolite MT23-GS16984

PESTANAL®, analytical standard

Synonym(s):

6-(tert-Butylamino)-1H-1,3,5-triazine-2,4-dione, Terbuthylazine-TP MT23 (LM5)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
HUF 21,100.00

HUF 21,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
HUF 21,100.00

About This Item

Empirical Formula (Hill Notation):
C7H12N4O2
CAS Number:
Molecular Weight:
184.20
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

HUF 21,100.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service