Skip to Content
Merck
All Photos(1)

Key Documents

67284-U

Supelco

BIOshell A160 Peptide C18 (2 µm) HPLC Columns

L × I.D. 5 cm × 1.5 mm, HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
HUF 419,000.00

HUF 419,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
HUF 419,000.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

HUF 419,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

BIOshell A160 Peptide C18 Column, 2 µm HPLC Column, 2.0 μm particle size, L × I.D. 5 cm × 1.5 mm

material

stainless steel hardware

Quality Level

Agency

suitable for USP L1

product line

BIOshell

feature

endcapped: no

analyte chemical class(es)

peptides

manufacturer/tradename

BIOshell

packaging

1 ea of

parameter

14,500 psi max. pressure (1000 bar)
60 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

5 cm × 1.5 mm

matrix

spherical silica particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

2.0 μm

pore size

160 Å pore size

operating pH range

1-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Shell thickness: 0.4 μm
Solid Core: 1.2 μm
Extent of labeling: 4.0%

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service