Skip to Content
Merck
All Photos(1)

Key Documents

48928

Supelco

ASTM® PS-18-44D Mix

in cyclohexane (varied conc.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® PS-18-44D

packaging

pkg of 6 × 1 mL

concentration

in cyclohexane (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Docosane 800 μg/mL
    Dotriacontane 1100 μg/mL
    Eicosane 500 μg/mL
    Hexacosane 1400 μg/mL
    Hexatriacontane 800 μg/mL
    Octacosane 1800 μg/mL
    Octadecane 300 μg/mL
    Tetracontane 500 μg/mL
    Tetracosane 1100 μg/mL
    Tetratetracontane 300 μg/mL
    Triacontane 1400 μg/mL
    See All (11)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service