Key Documents
22949
Supelco
Union, reducing
zero dead volume, for connecting 1/8 to 1/16 in., bore size 0.25 mm
Synonym(s):
Valco® Unions
Select a Size
1 EA
HUF 10,600.00
HUF 10,600.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
HUF 10,600.00
About This Item
UNSPSC Code:
41115700
HUF 10,600.00
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® IZR21C
bore size
0.25 mm
fitting
1/16 in. Valco (nut and ferrule included)
compatibility
for connecting 1/8 to 1/16 in.
General description
Stainless steel, unless noted otherwise. Nuts and ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2