Key Documents
22470-U
Supelco
Supelco Ferrule Nut Adapter (fits Agilent injectors and non-MS detectors), hexagonal wrenchtight version
pkg of 2 ea
Select a Size
2 EA
HUF 22,300.00
HUF 22,300.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
2 EA
HUF 22,300.00
About This Item
UNSPSC Code:
41121815
NACRES:
SB.54
HUF 22,300.00
Please contact Customer Service for Availability
Request a Bulk Order
General description
These stainless steel adapters enable you to use general purpose and other 1/16 in. compression ferrules in Agilent GCs. Hexagonal shape allows tightening with a wrench to keep fingers from getting burned.
Choose from one of the most recent versions:
Certificates of Analysis (COA)