Key Documents
Z374547
Sigma-Aldrich
Eppendorf® Pipet Helper®
Holds 0.1 to 100 mL pipettes
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
plastic
silicone
manufacturer/tradename
Eppendorf® 022230000
volume
0.1-100 mL
General description
Offers safe and convenient filling and dispensing with glass and plastic pipettes. Vacuum is applied by an easy-pull slide lever. The pipet helper also offers:
  • A single rocker-switch to accurately control aspiration and delivery
  • Disposable in-line filter to help prevent contamination
  • Holds 0.1 to 100 mL pipettes
Both filter and pipette connector are autoclavable and can be disassembled for maintenance and cleaning.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)