Key Documents
Z338699
VectaSpin Micro centrifuge tube filter
Ultrafiltration, pore size 20 kDa (molecular weight cut off), cellulose triacetate membrane
About This Item
Pricing and availability is not currently available.
material
cellulose triacetate membrane
polypropylene cap
polypropylene insert
sterility
non-sterile
packaging
pk of 100
manufacturer/tradename
Whatman 6834-2001
Whatman Article No. 28420452 (US reference)
pore size
20 kDa (molecular weight cut off)
General description
  • Quick and easy-to-use. Supplied ready assembled, saving time in the laboratory
  • Prefilter versions available for difficult-to-filter samples
  • Frosted area on tube for easy sample identification
  • 400μL insert capacity (receiving tube capacity with insert: 1.25mL; without insert: 2.0mL)
Applications:-
  • Removal of cells from culture media
  • Particle removal from solvents
  • Liquid chromatography sample preparation
  • Removal of bacteria from sample material
  • Fractionation/purification of proteins
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Choose from one of the most recent versions:
Certificates of Analysis (COA)