Skip to Content
Merck
All Photos(1)

Key Documents

W2895

Sigma-Aldrich

WE-14 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

Chromogranin A fragment 324-337 trifluoroacetate salt, H-Trp-Ser-Lys-Met-Asp-Gln-Leu-Ala-Lys-Glu-Leu-Thr-Ala-Glu-OH trifluoroacetate salt, Human CgA 324-337 trifluoroacetate salt, Peptide WE-14 trifluoroacetate salt, WSRMDQLAKELTAE trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H116N18O24S · xC2HF3O2
CAS Number:
Molecular Weight:
1649.86 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: >2 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CSCC[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CO)NC(=O)[C@@H](N)Cc1c[nH]c2ccccc12)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C72H116N18O24S.C2HF3O2/c1-35(2)29-50(67(108)78-37(5)59(100)80-44(17-11-13-26-73)62(103)82-47(20-23-55(94)95)65(106)87-51(30-36(3)4)69(110)90-58(39(7)92)71(112)79-38(6)60(101)85-49(72(113)114)21-24-56(96)97)86-64(105)46(19-22-54(76)93)83-68(109)52(32-57(98)99)88-66(107)48(25-28-115-8)84-63(104)45(18-12-14-27-74)81-70(111)53(34-91)89-61(102)42(75)31-40-33-77-43-16-10-9-15-41(40)43;3-2(4,5)1(6)7/h9-10,15-16,33,35-39,42,44-53,58,77,91-92H,11-14,17-32,34,73-75H2,1-8H3,(H2,76,93)(H,78,108)(H,79,112)(H,80,100)(H,81,111)(H,82,103)(H,83,109)(H,84,104)(H,85,101)(H,86,105)(H,87,106)(H,88,107)(H,89,102)(H,90,110)(H,94,95)(H,96,97)(H,98,99)(H,113,114);(H,6,7)/t37-,38-,39+,42-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,58-;/m0./s1

InChI key

FZKRZHJMJIARGO-YVXXVKOOSA-N

Biochem/physiol Actions

WE-14 is a Chromogranin A-derived peptide.
WE-14 peptide is a highly conserved neuropeptide derived from Chromogranin A (Parathyroid Secretory Protein 1). It has been detected in vertebrate and invertebrate neurons and in vertebrate endocrine tissues, most notably in the adrenal, pituitary, and parathyroid glands, and has also been found in human and bovine gastro-entero-pancreatic tissues.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colocalization of WE-14 immunostaining with the classical islet hormones in the porcine pancreas.
W J Curry et al.
Advances in experimental medicine and biology, 426, 139-144 (1997-01-01)
Chromogranin A and its derived peptides in the rat and porcine gastro-entero-pancreatic system. Expression, localization, and characterization.
W J Curry et al.
Advances in experimental medicine and biology, 482, 205-213 (2001-02-24)
P Norlén et al.
Regulatory peptides, 70(2-3), 121-133 (1997-06-18)
The ECL cells constitute the predominant endocrine cell population in the mucosa of the acid-secreting part of the stomach (fundus). They are rich in chromogranin A (CGA), histamine and histidine decarboxylase (HDC). They secrete CGA-derived peptides and histamine in response
W J Curry et al.
Regulatory peptides, 113(1-3), 41-47 (2003-04-11)
WE-14 is derived from the cell-specific posttranslational processing of chromogranin A (CgA) in subpopulations of neuroendocrine cells and neurons. Region- and site-specific chromogranin A, pancreastatin and WE-14 antisera were employed to study the generation of WE-14 in porcine ocular tissues.
S C Barkatullah et al.
Histochemistry and cell biology, 116(3), 255-262 (2001-10-31)
Immunohistochemical investigation of the post-translational processing of chromogranin A (CgA) to generate WE-14 in the sympathoadrenal cell lineage of the developing porcine fetus (F) detected intense CgA and weak WE-14 immunoreactivity in migrating neuroblast cells of the diffuse sympathetic ganglia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service