Skip to Content
Merck
All Photos(1)

Key Documents

T4050

Supelco

Trenbolone acetate

≥98%

Synonym(s):

17β-Acetoxyestra-4,9,11-trien-3-one

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product T4050 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥98%

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

General description

Trenbolone acetate is a synthetic anabolic steroid,[1] used as growth promoter in farm animals.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The fate of trenbolone acetate and melengestrol acetate after application as growth promoters in cattle: environmental studies.
Schiffer B, et al.
Environmental Health Perspectives, 109(11), 1145-1145 (2001)
Identification of metabolites of trenbolone acetate in androgenic runoff from a beef feedlot.
Durhan J E, et al.
Environmental Health Perspectives, 114(1), 65-65 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service