Skip to Content
Merck
All Photos(1)

Key Documents

SRP5183

Sigma-Aldrich

GAB1, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~108 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GAB1(2549)

General description

GAB1 is a member of the IRS1-like multisubstrate docking protein family and is a direct substrate of the epidermal growth factor receptor and the insulin receptor. GAB1 is tyrosine phosphorylated upon stimulation of cells with various cytokines, growth factors, and antigen receptor. Tyrosine phosphorylation of GAB1 mediates interaction with several proteins that contain SH2 domains such as SHP2 and phosphatidylinositol 3-kinase. GAB1 is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anatoly Kiyatkin et al.
The Journal of biological chemistry, 281(29), 19925-19938 (2006-05-12)
Grb2-associated binder 1 (GAB1) is a scaffold protein involved in numerous interactions that propagate signaling by growth factor and cytokine receptors. Here we explore in silico and validate in vivo the role of GAB1 in the control of mitogenic (Ras/MAPK)
M Holgado-Madruga et al.
Nature, 379(6565), 560-564 (1996-02-08)
The protein Grb2 plays a central role in signalling by receptor protein-tyrosine kinases, where its SH2 domain binds to the receptor and its two SH3 domains link to effectors. One target effector is Sos, so Grb2 links receptor protein-tyrosine kinases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service