Skip to Content
Merck
All Photos(1)

Key Documents

SML3848

Sigma-Aldrich

Y-320

≥98% (HPLC)

Synonym(s):

1-(4-Chlorophenyl)-N-[3-cyano-4-(4-morpholin-4-ylpiperidin-1-yl)phenyl]-5-methylpyrazole-4-carboxamide, 1-(4-Chlorophenyl)-N-[3-cyano-4-(4-morpholinopiperidin-1-yl)-phenyl]-5-methylpyrazole-4-carboxamide, 1-(4-Chlorophenyl)-N-[3-cyano-4-[4-(4-morpholinyl)-1-piperidinyl]phenyl]-5-methyl-1H-pyrazole-4-carboxamide, Y 320, Y320

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
HUF 35,200.00
50 MG
HUF 142,000.00

HUF 35,200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
HUF 35,200.00
50 MG
HUF 142,000.00

About This Item

Empirical Formula (Hill Notation):
C27H29ClN6O2
CAS Number:
Molecular Weight:
505.01
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

HUF 35,200.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N#CC1=CC(NC(C2=C(C)N(C3=CC=C(C=C3)Cl)N=C2)=O)=CC=C1N4CCC(CC4)N5CCOCC5

InChI

1S/C27H29ClN6O2/c1-19-25(18-30-34(19)24-5-2-21(28)3-6-24)27(35)31-22-4-7-26(20(16-22)17-29)33-10-8-23(9-11-33)32-12-14-36-15-13-32/h2-7,16,18,23H,8-15H2,1H3,(H,31,35)

InChI key

BWZNJVZTAWBIFG-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active immune modulator that suppresses inflammatory cytokines production and blocks P-glycoprotein-mediated MDR in vitro and in vivo.
Y-320 is an orally active immune modulator that inhibits IL-15-stimulated (100 ng/mL) IL-17 production from T-cells (IC50 = 57.4 nM using human CD4 T-cells & hIL-15; IC50 = 25.7/52.4 nM using murine CD4/Th17 T-cells and mIL-15 plus 1 μ/mL mCXCL12 & 1 μg/mL anti-CD3ε mAb) and prevents type II collagen-induced arthritis (CIA) in mice in vivo (0.3 to 3 mg/kg via daily p.o.). Y-320 is a P-glycoprotein (P-gp) substrate and sensitizes MDR tumor cells to chemotherapy drugs in cultures (paclitaxel IC50 with/without 500 nM Y-320 = 0.21/1.15 μM/Bads-200, 16/153 nM/Bads-72 breast cancer cells) and in mice in vivo (0% vs. 78% tumor growth suppression by 15 mg/kg PTX without or with 15 mg/kg Y-320 i.v.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y-320, a novel immune-modulator, sensitizes multidrug-resistant tumors to chemotherapy
American Journal of Translational Research, 12(2), 551-562 (2020)
Hiroyuki Ushio et al.
Pharmaceuticals (Basel, Switzerland), 7(1), 1-17 (2013-12-25)
Interleukin (IL)-15 and IL-17 are thought to play an important role in the pathogenesis of rheumatoid arthritis (RA) because both pro-inflammatory cytokines are found in synovial fluid of RA patients. In this study, we examined the pharmacological profiles of Y-320
Sara Hosseini-Farahabadi et al.
PLoS biology, 19(5), e3001221-e3001221 (2021-05-04)
Premature termination codons (PTC) cause over 10% of genetic disease cases. Some aminoglycosides that bind to the ribosome decoding center can induce PTC readthrough and restore low levels of full-length functional proteins. However, concomitant inhibition of protein synthesis limits the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service